missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-2-[5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine N-Oxide (Rizatriptan N-Oxide), Mikromol™
Description
N,N-Dimethyl-2-[5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine N-Oxide (Rizatriptan N-Oxide)
Specifications
Specifications
| Chemical Name or Material | Rizatriptan N10-Oxide |
| CAS | 260435-42-5 |
| Analyte or Component Names | Rizatriptan N10-Oxide |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C15 H19 N5 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Rizatriptan Benzoate Imp. H (EP), 1H-Indole-3-ethanamine, N,N-dimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-, N-oxide, N,N-Dimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indole-3-ethanamine N-oxide, Rizatriptan N-Oxide, Rizatriptan Benzoate Impurity H |
| InChI Formula | InChI=1S/C15H19N5O/c1-20(2,21)6-5-13-8-17-15-4-3-12(7-14(13)15)9-19-11-16-10-18-19/h3-4,7-8,10-11,17H,5-6,9H2,1-2H3 |
| SMILES | C[N+](C)([O-])CCc1c[nH]c2ccc(Cn3cncn3)cc12 |
| IUPAC Name | N,N-dimethyl-2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine oxide |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?