missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyldecylamine, TRC

Product Code. 30376550
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376550 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376550 Supplier Toronto Research Chemicals Supplier No. TRCD47230010ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Dimethyldecylamine
CAS 1120-24-7
Molecular Formula C12H27N
Quantity 10 mL
Synonym N,N-Dimethyl-1-decanamine,, N,N-Dimethyl-decylamine, 1-Dimethylaminodecane, ADMA 10, Armeen DM 10D, Decyldimethylamine, Dimethyldecylamine, Farmin DM 1098, Farmin DM 10P, N,N-Dimethyl-N-decylamine, N,N-Dimethyldecylamine, N-Decyl-N,N-dimethylamine, NSC 24529
InChI Formula InChI=1S/C12H27N/c1-4-5-6-7-8-9-10-11-12-13(2)3/h4-12H2,1-3H3
SMILES CN(CCCCCCCCCC)C
IUPAC Name N,N-dimethyldecan-1-amine
Molecular Weight (g/mol) 185.35
Formula Weight 185.2143
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.