Learn More
N,N-Dimethylsuccinamic acid, 98%
Product Code. 11322716
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322716 5 g 5g
11332716 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11322716 Supplier Thermo Scientific Alfa Aesar Supplier No. L08796.06
Request Bulk or Custom Format
CAS: 2564-95-6 | C6H11NO3 | 145.158 g/mol
It is employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2564-95-6
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.158
MDL Number MFCD00043505
InChI Key WAIGPJMZARQZDX-UHFFFAOYSA-N
Synonym n,n-dimethylsuccinamic acid, 4-dimethylamino-4-oxobutanoic acid, nn-dimethylsuccinamic acid, butanoic acid, 4-dimethylamino-4-oxo, n,n-dimethylsuccinic acid monoamide, 3-dimethylcarbamoyl propanoic acid, succinamic acid, n,n-dimethyl, n-dimethyl-succinamic acid, acmc-20ak89, succinamic acid,n-dimethyl
PubChem CID 75717
IUPAC Name 4-(dimethylamino)-4-oxobutanoic acid
SMILES CN(C)C(=O)CCC(=O)O
Specifications
Melting Point 82°C to 87°C
Quantity 5 g
Beilstein 1762788
Solubility Information Slightly soluble in water.
Formula Weight 145.16
Percent Purity 98%
Chemical Name or Material N,N-Dimethylsuccinamic acid