Learn More
N,N'-Methylene Ambroxol Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | trans-4-(6,8-Dibromo-1,4-dihydroquinazolin-3(2H)-yl)cyclohexanol Hydrochloride |
| CAS | 15942-08-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14 H18 Br2 N2 O . Cl H |
| Quantity | 500 mg |
| Synonym | trans-4-(6,8-Dibromo-1,4-dihydroquinazolin-3(2H)-yl)cyclohexanol Hydrochloride, Ambroxol Hydrochloride Imp. B (EP) as Hydrochloride, trans-4-(6,8-Dibromo-1,4-dihydro-3(2H)-quinazolinyl)cyclohexanol monohydrochloride |
| InChI Formula | InChI=1S/C14H18Br2N2O.ClH/c15-10-5-9-7-18(8-17-14(9)13(16)6-10)11-1-3-12(19)4-2-11;/h5-6,11-12,17,19H,1-4,7-8H2;1H/t11-,12-; |
| SMILES | Cl.O[C@@H]1CC[C@H](CC1)N2CNc3c(Br)cc(Br)cc3C2 |
| IUPAC Name | 4-(6,8-dibromo-2,4-dihydro-1H-quinazolin-3-yl)cyclohexan-1-ol;hydrochloride |
| Molecular Weight (g/mol) | 426.57 |
| Show More |