Learn More
N-(n-Propyl)maleimide, 94%, Thermo Scientific™
Product Code. 11300054
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11300054
Brand: Thermo Scientific Alfa Aesar L00460.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21746-40-7
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.15
MDL Number MFCD00014541
InChI Key DABFKTHTXOELJF-UHFFFAOYSA-N
Synonym n-propylmaleimide, 1-propyl-1h-pyrrole-2,5-dione, n-n-propyl maleimide, n-propyl maleimide, n-n-propylmaleimide, 1h-pyrrole-2,5-dione, 1-propyl, 1-propyl-2,5-dihydro-1h-pyrrole-2,5-dione, n-propylmaleinimide, acmc-20ak14
PubChem CID 89033
SMILES CCCN1C(=O)C=CC1=O
Specifications
Melting Point 24°C to 26°C
Density 1.073
Boiling Point 97°C (18 mmHg)
Flash Point 88°C (190°F)
Refractive Index 1.48
Quantity 5 g
UN Number UN2811
Beilstein 1525008
Solubility Information Insoluble in water.
IUPAC Name 1-propylpyrrole-2,5-dione
Formula Weight 139.15
Percent Purity 94%
Chemical Name or Material N-(n-Propyl)maleimide