missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso Azithromycin, TRC

Product Code. 30480951
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480951 5 mg 5mg
30465034 1 mg 1mg
30483853 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480951 Supplier Toronto Research Chemicals Supplier No. TRCN1374785MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitroso Azithromycin
Analyte or Component Names N-Nitroso Azithromycin
Shipping Condition Room Temperature
Molecular Formula C37 H69 N3 O13
Synonym (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-(((2S,3R,4S,6R)-4-(Dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2-ethyl-3,4,10-trihydroxy-13-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,8,10,12,14-hexamethyl-6-nitroso-1-oxa-6-azacyclopentadecan-15-one
InChI Formula InChI=1S/C37H69N3O13/c1-14-26-37(10,46)30(42)23(6)40(38-47)18-19(2)16-35(8,45)32(53-34-28(41)25(39(11)12)15-20(3)49-34)21(4)29(22(5)33(44)51-26)52-27-17-36(9,48-13)31(43)24(7)50-27/h19-32,34,41-43,45-46H,14-18H2,1-13H3/t19-,20-,21+,22-,23-,24+,25+,26-,27+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1
Solution Type Neat
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN(N=O)[C@H](C)[C@@H](O)[C@]1(C)O
IUPAC Name (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-2-ethyl-3,4,10-trihydroxy-13-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,8,10,12,14-hexamethyl-6-nitroso-1-oxa-6-azacyclopentadecan-15-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 763.956
Quantity 5 mg
Formula Weight 763.483
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.