Learn More
N-Nitroso Duloxetine, TRC
Product Code. 30007588
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007588 250 mg 250mg
30006777 500 mg 500mg
17901285 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30007588 Supplier Toronto Research Chemicals Supplier No. TRCN791860250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2680527-91-5
Molecular Formula C18 H18 N2 O2 S
Molecular Weight (g/mol) 326.413
Synonym 2-Thiophenepropanamine, N-methyl-γ-(1-naphthalenyloxy)-N-nitroso- (ACI), N-Methyl-N-[3-(1-naphthyloxy)-3-thien-2-ylpropyl]-N-nitrosoamine, N-Methyl-γ-(1-naphthalenyloxy)-N-nitroso-2-thiophenepropanamine, N-Nitroso-(±)-duloxetine, rac-N-Nitrosoduloxetine
IUPAC Name N-methyl-N-(3-naphthalen-1-yloxy-3-thiophen-2-ylpropyl)nitrous amide
SMILES CN(CCC(Oc1cccc2ccccc12)c3cccs3)N=O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C18H18N2O2S/c1-20(19-21)12-11-17(18-10-5-13-23-18)22-16-9-4-7-14-6-2-3-8-15(14)16/h2-10,13,17H,11-12H2,1H3
Formula Weight 326.109
Percent Purity >95%
Chemical Name or Material (+/-)-N-Nitrosoduloxetine
Grade HPLC