missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitroso-N-isopropyl-4-aminobutyric Acid, Mikromol™
Description
N-Nitroso-N-isopropyl-4-aminobutyric Acid
Specifications
Specifications
| Chemical Name or Material | N-Nitroso-N-isopropyl-4-aminobutyric Acid |
| CAS | 2624122-60-5 |
| Analyte or Component Names | N-Nitroso-N-isopropyl-4-aminobutyric Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H14 N2 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-Nitroso-N-isopropyl-4-aminobutyric Acid, 4-[(1-Methylethyl)nitrosoamino]butanoic acid, Butanoic acid, 4-[(1-methylethyl)nitrosoamino]- |
| InChI Formula | InChI=1S/C7H14N2O3/c1-6(2)9(8-12)5-3-4-7(10)11/h6H,3-5H2,1-2H3,(H,10,11) |
| SMILES | CC(C)N(CCCC(=O)O)N=O |
| IUPAC Name | 4-[nitroso(propan-2-yl)amino]butanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?