Learn More
N-Nitrosomorpholine-d8, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Nitrosomorpholine-d8 |
| CAS | 1219805-76-1 |
| Molecular Formula | C4 2H8 N2 O2 |
| Quantity | 0.05 g |
| Synonym | 2,2,3,3,5,5,6,6-Octadeutero-N-nitrosomorpholine, 4-Nitrosomorpholine-2,2,3,3,5,5,6,6-d8, 4-Nitrosomorpholine D8, N-Nitrosomorpholine-2,2,3,3,5,5,6,6-d8, N-Nitrosomorpholine D8 |
| InChI Formula | InChI=1 S/C4H8N2O2/c7-5-6-1-3-8-4-2-6/h1-4H2/i1D2,2D2,3D2,4D2 |
| SMILES | [2 H]C1([2 H])OC([2 H])([2 H])C([2 H])([2 H])N(N=O)C1([2 H])[2 H] |
| IUPAC Name | 2,2,3,3,5,5,6,6-octadeuterio-4-nitrosomorpholine |
| Molecular Weight (g/mol) | 124.17 |
| Formula Weight | 124.1088 g/mol |
| Show More |