missing translation for 'onlineSavingsMsg'
Learn More

n-Nonadecanoic Acid, TRC

Product Code. 30483561
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Product Code. Quantity
30483561 5 g
30468455 10 g
30473101 1 g
3 options
This item is not returnable. View return policy

Product Code. 30483561

Brand: Toronto Research Chemicals undefined

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-30-0
Formula Weight 298.2872
InChI Formula InChI=1S/C19H38O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21/h2-18H2,1H3,(H,20,21)
IUPAC Name nonadecanoic acid
Molecular Formula C19 H38 O2
Molecular Weight (g/mol) 298.5
SMILES CCCCCCCCCCCCCCCCCCC(=O)O
Synonym Nonadecanoic acid (6CI, 8CI, 9CI, ACI), n-Nonadecanoic acid, n-Nonadecylic acid, Nonadecylic acid, NSC 11914
Chemical Name or Material Nonadecanoic acid (Nonadecyclic acid)

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.