missing translation for 'onlineSavingsMsg'
Learn More

N-(p-Toluenesulfonyl)-L-phenylalanine chloromethyl ketone, Thermo Scientific™

Catalog Number p-7058187
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
402-71-1
C17H18ClNO3S
351.85
MFCD00000935
MQUQNUAYKLCRME-INIZCTEOSA-N
tpck, tos-phe-ch2cl, tosylphenylalanyl chloromethyl ketone, n-tosyl-l-phenylalanyl chloromethyl ketone, n-tosyl-l-phenylalanine chloromethyl ketone, n-p-tosyl-l-phenylalanine chloromethyl ketone, l-1-tosylamido-2-phenylethyl chloromethyl ketone, l-n-alpha-chloroacetyl phenethyl-p-toluenesulfonamide, tos-phe-chloromethylketone, chembl60718
439647
CHEBI:9642
CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CC1=CC=CC=C1)C(=O)CCl
This item is not returnable. View return policy
402-71-1
C17H18ClNO3S
351.85
MFCD00000935
MQUQNUAYKLCRME-INIZCTEOSA-N
tpck, tos-phe-ch2cl, tosylphenylalanyl chloromethyl ketone, n-tosyl-l-phenylalanyl chloromethyl ketone, n-tosyl-l-phenylalanine chloromethyl ketone, n-p-tosyl-l-phenylalanine chloromethyl ketone, l-1-tosylamido-2-phenylethyl chloromethyl ketone, l-n-alpha-chloroacetyl phenethyl-p-toluenesulfonamide, tos-phe-chloromethylketone, chembl60718
439647
CHEBI:9642
CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CC1=CC=CC=C1)C(=O)CCl

Enzyme inhibitor that inactivates chymotrypsin but not trypsin

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Beilstein 2895215
Solubility Information Soluble in DMSO at 10mg/ml
IUPAC Name N-[(2S)-4-chloro-3-oxo-1-phenylbutan-2-yl]-4-methylbenzenesulfonamide
Formula Weight 351.85
Quantity 1 g
Physical Form Powder
Chemical Name or Material N-(p-Toluenesulfonyl)-L-phenylalanine chloromethyl ketone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'