missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-2-naphthylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-4813587
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
135-88-6
C16H13N
219.29
MFCD00004052
KEQFTVQCIQJIQW-UHFFFAOYSA-N
n-phenyl-2-naphthylamine, n-2-naphthyl aniline, neozone, aceto pbn, stabilizator ar, agerite powder, anilinonaphthalene, antioxidant 116, pbna, 2-naphthylphenylamine
8679
CHEBI:34877
N-phenylnaphthalen-2-amine
N(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
This item is not returnable. View return policy
135-88-6
C16H13N
219.29
MFCD00004052
KEQFTVQCIQJIQW-UHFFFAOYSA-N
n-phenyl-2-naphthylamine, n-2-naphthyl aniline, neozone, aceto pbn, stabilizator ar, agerite powder, anilinonaphthalene, antioxidant 116, pbna, 2-naphthylphenylamine
8679
CHEBI:34877
N-phenylnaphthalen-2-amine
N(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 105°C to 108°C
Boiling Point 395°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene,acetone,acetic acid
Formula Weight 219.28
Percent Purity 97%
Chemical Name or Material N-Phenyl-2-naphthylamine
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Phenyl-2-naphthylamine
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'