missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-N-(4-piperidinyl)propionamide, 98%, Thermo Scientific™

Product Code. 11461790
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11461790

Brand: Thermo Scientific Alfa Aesar H27522.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Norfentanyl is a major urinary metabolite of a narcotic analgesic fentanyl.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1609-66-1
Molecular Formula C14H20N2O
Molecular Weight (g/mol) 232.327
MDL Number MFCD00145005
InChI Key PMCBDBWCQQBSRJ-UHFFFAOYSA-N
Synonym norfentanyl, n-phenyl-n-piperidin-4-yl propionamide, n-phenyl-n-piperidin-4-yl-propionamide, 4-n-propionylanilino piperidine, unii-2mk6d8jv6j, n-phenyl-n-4-piperidinyl propanamide, propanamide, n-phenyl-n-4-piperidinyl, n-phenyl-n-4-piperidinylpropanamide, n-phenyl-n-piperidin-4-yl propanamide, n-phenyl-n-4-piperidinylpropionamide
PubChem CID 259381
ChEBI CHEBI:62685
IUPAC Name N-phenyl-N-piperidin-4-ylpropanamide
SMILES CCC(=O)N(C1CCNCC1)C2=CC=CC=C2

Specifications

Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 232.33
Percent Purity 98%
Chemical Name or Material N-Phenyl-N-(4-piperidinyl)propionamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.