Learn More
n-Propyl-d7 Alcohol, CDN
Product Code. 30319557
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319557 1 g 1g
30322208 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319557 Supplier CDN Supplier No. CDND12361G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Propanol D7
CAS 102910-31-6
Molecular Formula C3 2H7 H O
Quantity 1 g
Synonym 1-Propanol D7, 1,1,2,2,3,3,3-Heptadeuteriopropan-1-ol, 1-Propan-1,1,2,2,3,3,3-d7-ol (9 CI)
InChI Formula InChI=1 S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3/i1D3,2D2,3D2
SMILES [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O
IUPAC Name 1,1,2,2,3,3,3-heptadeuteriopropan-1-ol
Molecular Weight (g/mol) 67.14
Formula Weight 67.1015 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity