Learn More
Invitrogen™ N-terminal Arginylation Monoclonal Antibody (4A9), FITC
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545371
Product Code. 17878662
  • € 481.00 / 100µg
Description
A 1:1,000 dilution of MA5-45371 was sufficient for detection of N-terminal Arginylation in 0.5 μg of N-terminal Arginine peptide conjugated to BSA by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Specific for N-terminal arginine next to glutamic acid. Does not detect N-terminal arginine next to aspartic acid or internal arginine residues.
Protein arginylation is the post-translational addition of arginine to proteins by arginyltransferase ATE1. Arginylation of proteins has been found to play an essential role in physiological pathways during embryogenesis and adulthood. Arginylation has also been shown to regulate cell stress responses, including ER stress, cytosolic misfolded proteins, and heat stress.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
N-terminal Arginylation
Monoclonal
1 mg/mL
PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Mouse
Protein G
RUO
Chemical
Antibody
IgG1
Flow Cytometry, Western Blot
4A9
FITC
N terminal Arginine; N-terminal Arginine
Synthetic N-terminal arginylated peptide conjugated to KLH.
100 μg
Primary
4°C, store in dark
Liquid
Chemical