missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ N-WASP Monoclonal Antibody (C.427.3)
Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA515011
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
The Wiskott-Aldrich syndrome (WAS) family of proteins share similar domain structure, and are involved in transduction of signals from receptors on the cell surface to the actin cytoskeleton. The presence of a number of different motifs suggests that they are regulated by a number of different stimuli, and interact with multiple proteins. Recent studies have demonstrated that these proteins, directly or indirectly, associate with the small GTPase, Cdc42, known to regulate formation of actin filaments, and the cytoskeletal organizing complex, Arp2/3. The WASL gene product is a homolog of WAS protein, however, unlike the latter, it is ubiquitously expressed and shows highest expression in neural tissues. It has been shown to bind Cdc42 directly, and induce formation of long actin microspikes.
Specifications
| N-WASP | |
| Monoclonal | |
| 8 μg/mL | |
| 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| O00401, O08816, Q91YD9, Q95107 | |
| WASL | |
| Synthetic peptide corresponding the sequence of human N-WASP. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Bovine, Hamster | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| C.427.3 | |
| Unconjugated | |
| WASL | |
| 2900021I12Rik; 3110031I02Rik; DKFZp779G0847; MGC48327; n wasp; neural Wiskott-Aldrich syndrome protein; NWASP; N-WASP; TRS4; WASL; WASP like actin nucleation promoting factor; WASPB; Wiskott-Aldrich syndrome like; Wiskott-Aldrich syndrome-like; Wiskott-Aldrich syndrome-like (human) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100757697, 281577, 682507, 73178, 8976 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction