Learn More
Invitrogen™ N-WASP Recombinant Rabbit Monoclonal Antibody (23GB2850), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552529
 View more versions of this product
Product Code. 30087450
  • € 186.00 / 20µL
Description
The Wiskott-Aldrich syndrome (WAS) family of proteins share similar domain structure, and are involved in transduction of signals from receptors on the cell surface to the actin cytoskeleton. The presence of a number of different motifs suggests that they are regulated by a number of different stimuli, and interact with multiple proteins. Recent studies have demonstrated that these proteins, directly or indirectly, associate with the small GTPase, Cdc42, known to regulate formation of actin filaments, and the cytoskeletal organizing complex, Arp2/3. The WASL gene product is a homolog of WAS protein, however, unlike the latter, it is ubiquitously expressed and shows highest expression in neural tissues. It has been shown to bind Cdc42 directly, and induce formation of long actin microspikes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
N-WASP
Recombinant Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O00401, O08816, Q91YD9
WASL
A synthesized peptide derived from human WASL (400-450AA).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB2850
Unconjugated
WASL
2900021I12Rik; 3110031I02Rik; DKFZp779G0847; MGC48327; n wasp; neural Wiskott-Aldrich syndrome protein; NWASP; N-WASP; TRS4; WASL; WASP like actin nucleation promoting factor; WASPB; Wiskott-Aldrich syndrome like; Wiskott-Aldrich syndrome-like; Wiskott-Aldrich syndrome-like (human)
Rabbit
Affinity chromatography
RUO
682507, 73178, 8976
-20°C