Learn More
N1-((Ethylimino)methylene)-N3,N3-dimethylpropane-1,3-diamine, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 1892-57-5 |
| Molecular Formula | C8H17N3 |
| MDL Number | MFCD00044916 |
| Quantity | 10 g |
| SMILES | CN(C)CCCN=C=NCC |
| Molecular Weight (g/mol) | 155.24 |
| Percent Purity | 98% |
| Chemical Name or Material | N1-((Ethylimino)methylene)-N3,N3-dimethylpropane-1,3-diamine |