Learn More
N2-(Benzyloxycarbonyl)-N6-(tert-butoxycarbonyl)-L-lysine, Mikromol™
Description
N2-(Benzyloxycarbonyl)-N6-(tert-butoxycarbonyl)-L-lysine
Specifications
Specifications
| Chemical Name or Material | N2-(Benzyloxycarbonyl)-N6-(tert-butoxycarbonyl)-L-lysine |
| CAS | 2389-60-8 |
| Analyte or Component Names | N2-(Benzyloxycarbonyl)-N6-(tert-butoxycarbonyl)-L-lysine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H28 N2 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-Lysine, N6-[(1,1-dimethylethoxy)carbonyl]-N2-[(phenylmethoxy)carbonyl]-, Lysine, N2,N6-dicarboxy-, N2-benzyl N6-tert-butyl ester, L- (7CI,8CI), N6-[(1,1-Dimethylethoxy)carbonyl]-N2-[(phenylmethoxy)carbonyl]-L-lysine, (S)-2-(Benzyloxycarbonylamino)-6-(tert-butoxycarbonylamino)hexanoic acid, Cbz-Lys(Boc)-OH, N-α-Benzyloxycarbonyl-N-ε-tert-butoxycarbonyl-L-lysine, N2-Benzyloxycarbonyl-N6-tert-butoxycarbonyl-L-lysine, NSC 164047, Nε-[(tert-Butoxy)carbonyl]-Nα--[(benzyloxy)carbonyl]-L-lysine, Z-Lys(Boc)-OH |
| InChI Formula | InChI=1S/C19H28N2O6/c1-19(2,3)27-17(24)20-12-8-7-11-15(16(22)23)21-18(25)26-13-14-9-5-4-6-10-14/h4-6,9-10,15H,7-8,11-13H2,1-3H3,(H,20,24)(H,21,25)(H,22,23)/t15-/m0/s1 |
| SMILES | CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCc1ccccc1)C(=O)O |
| IUPAC Name | (2S)-6-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)hexanoic acid |
| Show More |