missing translation for 'onlineSavingsMsg'
Learn More

N2-Trityl Olmesartan Acid, TRC

Product Code. 30450814
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450814 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450814 Supplier Toronto Research Chemicals Supplier No. TRCT8894105MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N2-Tritylolmesartan
CAS 752179-89-8
Molecular Formula C43 H40 N6 O3
Quantity 5 mg
Synonym 1H-Imidazole-5-carboxylic acid','4-(1-hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-, 4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic acid
InChI Formula InChI=1S/C43H40N6O3/c1-4-16-37-44-39(42(2,3)52)38(41(50)51)48(37)29-30-25-27-31(28-26-30)35-23-14-15-24-36(35)40-45-47-49(46-40)43(32-17-8-5-9-18-32,33-19-10-6-11-20-33)34-21-12-7-13-22-34/h5-15,17-28,52H,4,16,29H2,1-3H3,(H,50,51)
SMILES CCCc1nc(c(C(=O)O)n1Cc2ccc(cc2)c3ccccc3c4nnn(n4)C(c5ccccc5)(c6ccccc6)c7ccccc7)C(C)(C)O
IUPAC Name 5-(2-hydroxypropan-2-yl)-2-propyl-3-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylic acid
Molecular Weight (g/mol) 688.82
Formula Weight 688.3162
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.