missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N4BP2L2 Antibody (OTI1C8), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72866AF647
This item is not returnable.
View return policy
Description
N4BP2L2 Monoclonal specifically detects N4BP2L2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| N4BP2L2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| N4BP2L2 | |
| Full length recombinant protein of human N4BP2L2 (NP_055702) produced in E.coli. | |
| 0.1 mL | |
| 10443.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1C8 | |
| Western Blot, Immunohistochemistry | |
| CG005FLJ41089, CG016, FLJ36195, NEDD4 binding protein 2-like 2, NEDD4-binding protein 2-like 2, PFAAP5FLJ43077, Phosphonoformate immuno-associated protein 5,92M18.3,92M18.3 (novel protein), protein from BCRA2 region | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction