Learn More
ZNF264 Monoclonal Antibody (OTI8G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810500
Encompass_Preferred
 View more versions of this product
Product Code. 17563644
  • € 540.87 / 100µL
Description
ZNF264 gene encodes a zinc finger protein and belongs to the krueppel C2H2-type zinc-finger protein family. Zinc finger proteins are often localized in the nucleus, bind nucleic acids, and regulate transcription.
Specifications
Specifications
ZNF264
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O43296
ZNF264
Full length human recombit protein of human ZNF264 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI8G5
Unconjugated
ZNF264
zinc finger protein 264
Mouse
Affinity Chromatography
RUO
9422
-20° C, Avoid Freeze/Thaw Cycles