missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ N6-Cyclohexyladenosine

Product Code. 18798706
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18798706

Brand: Tocris Bioscience™ 6863/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High affinity A1 receptor agonist

N6-Cyclohexyladenosine is a high affinity adenosine A1 receptor agonist (Kd values are 0.7 and 6 nM for bovine and guinea pig brain membranes, respectively). Reduces light-induced circadian phase delays. Exhibits protective effect in lysolecithin-induced demyelination model. When administered prior to ischemia, improves functional recovery of heart in a mouse model of ischemia-reperfusion injury. Induces hypothermia following icv administration in mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Cyclohexyladenosine
CAS 36396-99-3
Quantity 50 mg
Molecular Formula C16H23N5O4
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.