missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-methyladenosine (m6A) Antibody (17-3-4-1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-50525CL1
This item is not returnable.
View return policy
Description
N6-methyladenosine (m6A) Monoclonal antibody specifically detects N6-methyladenosine (m6A) in Human,Mouse,Yeast samples. It is validated for Immunoprecipitation,Western Blot,Dot Blot
Specifications
| N6-methyladenosine (m6A) | |
| Monoclonal | |
| CoraFluor 1 | |
| (-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine | |
| Hapten N6-methyladenosine-5'-mono-phosphate conjugated to BSA of all N6-methyladenosine | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 λ |
| Immunoprecipitation, Western Blot, Dot Blot | |
| 17-3-4-1 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human, Mouse, Yeast | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction