missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-methyladenosine (m6A) Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
€ 214.00 - € 551.00
Specifications
| Antigen | N6-methyladenosine (m6A) |
|---|---|
| Dilution | ELISA Recommended starting concentration is 1 μg/mL, Dot Blot 1:500 - 1:2000 |
| Applications | ELISA, Dot Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230665
|
Novus Biologicals
NBP3-33194-100ul |
100 μL |
€ 551.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232175
|
Novus Biologicals
NBP3-33194-20ul |
20 μL |
€ 214.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N6-methyladenosine (m6A) Monoclonal antibody specifically detects N6-methyladenosine (m6A) in Nonspecies specific samples. It is validated for ELISA,Dot BlotSpecifications
| N6-methyladenosine (m6A) | |
| ELISA, Dot Blot | |
| Unconjugated | |
| Mouse | |
| Non-species specific | |
| (-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine | |
| IgG1 κ | |
| Affinity purified |
| ELISA Recommended starting concentration is 1 μg/mL, Dot Blot 1:500 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| Chemical compounds corresponding to N6-methyladenosine (m6A). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title