missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ N6-Methyladenosine (m6A) Monoclonal Antibody (A8F2)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA544938

Product Code. 17820356

  • € 494.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Application notes: Indirect ELISA.

N6-methyladenosine (m6A), or methylation of the N6 position of adenosine is a posttranscriptional modification of RNA. Due to a lack of analytical methods, N6-methyladenosine is poorly understood, but new evidence suggests that it is a very common base modification and important physiological regulator. N6-methyladenosine is markedly increased throughout brain development, and is enriched near stop codons, microRNA-binding sites and UTRs, which indicates a fundamental role in the regulation of gene expression. N6-methyladenosine is also highly conserved between human and mouse.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

N6-Methyladenosine (m6A)
Monoclonal
2 mg/mL
PBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Mouse
Protein A
RUO
Chemical
Antibody
IgG1
Dot Blot, ELISA
A8F2
Unconjugated
C11H15N5O4; m6A
m6A Small Molecule conjugated to OVA.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Chemical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.