missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ N6-Methyladenosine (m6A) Recombinant Rabbit Monoclonal Antibody (6S5W2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535350
This item is not returnable.
View return policy
Description
N6-Methyladenosine (m6A) Recombinant Monoclonal Antibody for DB, ELISA, ICC/IF, RIP
N6-methyladenosine (m6A), or methylation of the N6 position of adenosine is a posttranscriptional modification of RNA. Due to a lack of analytical methods, N6-methyladenosine is poorly understood, but new evidence suggests that it is a very common base modification and important physiological regulator. N6-methyladenosine is markedly increased throughout brain development, and is enriched near stop codons, microRNA-binding sites and UTRs, which indicates a fundamental role in the regulation of gene expression. N6-methyladenosine is also highly conserved between human and mouse.
Specifications
| N6-Methyladenosine (m6A) | |
| Recombinant Monoclonal | |
| 1.05 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.05% ProClin 300; pH 7.3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Chemical, Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Dot Blot, ELISA, RNA Immunoprecipitation, Immunocytochemistry | |
| 6S5W2 | |
| Unconjugated | |
| C11H15N5O4; m6A | |
| Chemical compounds corresponding to N6-methyladenosine / m6A. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction