missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ N6AMT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566242
This item is not returnable.
View return policy
Description
Immunogen sequence: GREVMDRFFPL VPDLLSPRGL FYLVTIKENN PEEILKIMKT KGLQGTTALS RQAGQETLSV LKFT Highest antigen sequence identity to the following orthologs - mouse 85%, rat 88%.
Heterodimeric methyltransferase that catalyzes N5-methylation of ETF1 on 'Gln-185', using S-adenosyl L-methionine as methyl donor. ETF1 needs to be complexed to ERF3 in its GTP-bound form to be efficiently methylated. May play a role in the modulation of arsenic-induced toxicity. May be involved in the conversion of monomethylarsonous acid (3+) into the less toxic dimethylarsonic acid.
Specifications
| N6AMT1 | |
| Polyclonal | |
| Unconjugated | |
| N6AMT1 | |
| 5830445C04Rik; C21orf127; HemK methyltransferase family member 2; HEMK2; m.HsaHemK2P; Methylarsonite methyltransferase N6AMT1; Methyltransferase N6AMT1; methyltransferase N6AMT1; hemK methyltransferase family member 2; M-N6DNA MTase A; MTQ2; N(6)-adenine-specific DNA methyltransferase 1; N(6)-adenine-specific DNA methyltransferase 1-like; N-6 adenine-specific DNA methyltransferase 1; N-6 adenine-specific DNA methyltransferase 1 (putative); N6AMT; N6amt1; N6-DNA methyltransferase A; N6-DNA-methyltransferase; PRED28; Protein N(5)-glutamine methyltransferase; RGD1311843 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29104 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y5N5 | |
| N6AMT1 | |
| Recombinant Human N6AMT1. Recombinant protein control fragment (Product #RP-106922). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction