missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAA15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530562
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (93%), Rat (92%), Rhesus Monkey (97%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein of unknown function. However, similarity to proteins in yeast and other species suggests that this protein may be an N-acetyltransferase.
Specifications
| NAA15 | |
| Polyclonal | |
| Unconjugated | |
| NAA15 | |
| 5730450D16Rik; 6330400I15; ASTBDN; GA19; gastric cancer antigen Ga19; mNAT1; N(alpha)-acetyltransferase 15, NatA auxiliary subunit; Naa15; N-alpha-acetyltransferase 15, NatA auxiliary subunit; NARG1; NAT1P; NATH; NMDA receptor regulated 1; NMDA receptor-regulated gene 1; NMDA receptor-regulated protein 1; N-terminal acetyltransferase; N-terminal acetyltransferase 1; N-terminal aceyltransferase 1; Protein tubedown-1; TBDN; Tbdn-1; TBDN100; transcriptional coactivator tubedown-100; tubedown; tubedown-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80155 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9BXJ9 | |
| NAA15 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human NARG1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction