missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAA38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113373
This item is not returnable.
View return policy
Description
NAA38 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
NAA38 is a protein coding gene.
Specifications
| NAA38 | |
| Polyclonal | |
| Unconjugated | |
| NAA38 | |
| 1500034E06Rik; fj33f12; LSM domain containing 1; LSM domain-containing protein 1; LSMD1; MAK31; MAK31-like protein; N(alpha)-acetyltransferase 38, NatC auxiliary subunit; naa38; N-alpha-acetyltransferase 38, NatC auxiliary subunit; PFAAP2; Phosphonoformate immuno-associated protein 2; RGD1559617; si:ch211-30g3.3; wu:fj33f12 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 78304, 84316 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9BRA0, Q9D2U5 | |
| NAA38 | |
| Recombinant Human N-alpha-acetyltransferase 38, NatC auxiliary subunit protein (2-173AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction