Learn More
Invitrogen™ NAA40 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140084
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: KSSKAKEKKQ KRLEERAAMD AVCAKVDAAN RLGDPLEAFP VFKKYDRNGL NVSIECKRVS GLEPATVDWA FD Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
N-terminal acetylation is one of the most common protein modifications in eukaryotes, occurring on approximately 57% and 84% on yeast and human proteins respectively. There are several N-terminal acetylating enzyme complexes (NatA - NatE). Unlike the other complexes, NatD is composed of a single protein, NAT11, and has recently been described to acetylate the Serine N-termini of histones H2A and H4 in yeast. The role these modifications play is unknown; yeast that do not express NAT11 grow at normal rates and have no observable phenotypes. The role of the human homolog is likewise unknown.
Specifications
| NAA40 | |
| Polyclonal | |
| Unconjugated | |
| NAA40 | |
| 4931433E08Rik; AU023197; N(alpha)-acetyltransferase 40, NatD catalytic subunit; N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog; N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog (S. cerevisiae); Naa40; N-acetyltransferase 11; N-acetyltransferase 11 (GCN5-related, putative); N-alpha acetyl transferase 40; Nalpha acetyltransferase 40; N-alpha-acetyltransferase 40; N-alpha-acetyltransferase 40, NatD catalytic subunit; N-alpha-acetyltransferase D; NAT11; NatD; natD catalytic subunit; PATT1; Protein acetyltransferase 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79829 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86UY6 | |
| NAA40 | |
| Recombinant protein corresponding to Human NAA40. Recombinant protein control fragment (Product #RP-108926). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.