missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAALADase-like 1/NAALADL1 Antibody (29), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89819MFV450
This item is not returnable.
View return policy
Description
NAALADase-like 1/NAALADL1 Monoclonal antibody specifically detects NAALADase-like 1/NAALADL1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| NAALADase-like 1/NAALADL1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 10004 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 29 | |
| Western Blot, ELISA | |
| I100, NAALADASEL, NAALADL1 N-acetylated alpha-linked acidic dipeptidase-like 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human NAALADase-like 1/NAALADL1 (Accession#: NP_005459.2; Pro 29-Leu 740). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction