missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAB1 Antibody (OTI1D2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72867F
This item is not returnable.
View return policy
Description
NAB1 Monoclonal specifically detects NAB1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| NAB1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| NAB1 | |
| Recombinant protein fragment corresponding to amino acids 237-487 of human NAB1 (NP_005957) produced in E.coli. | |
| 0.1 mL | |
| 4664.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI1D2 | |
| Immunohistochemistry | |
| EGR1 binding protein 1, EGR-1-binding protein 1, NGFI-A binding protein 1 (EGR1 binding protein 1), NGFI-A-binding protein 1, Transcriptional regulatory protein p54 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction