missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAB1 Monoclonal Antibody (OTI5B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802910S
This item is not returnable.
View return policy
Description
NAB1 Monoclonal antibody specifically detects NAB1 in Human samples. It is validated for Western BlotSpecifications
| NAB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13506 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4664 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI5B11 | |
| Unconjugated | |
| NAB1 | |
| EGR1 binding protein 1; EGR-1-binding protein 1; Nab1; NGF1A-binding protein; Ngfi-A binding protein 1; NGFI-A binding protein 1 (EGR1 binding protein 1); NGFI-A-binding protein 1; Transcriptional regulatory protein p54 | |
| Human recombinant protein fragment corresponding to amino acids 237-487 of human NAB1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction