missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nab2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99585F
This item is not returnable.
View return policy
Description
Nab2 Polyclonal antibody specifically detects Nab2 in Human samples. It is validated for Western Blot
Specifications
| Nab2 | |
| Polyclonal | |
| Western Blot | |
| EGR-1-binding protein 2, MADEREGR1 binding protein 2, Melanoma-associated delayed early response protein, MGC75085, NGFI-A binding protein 2 (EGR1 binding protein 2), NGFI-A-binding protein 2, Protein MADER | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Nab2. | |
| 0.1 mL | |
| Neuroscience, Transcription Factors and Regulators | |
| 4665 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| FITC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction