missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nab2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19463
This item is not returnable.
View return policy
Description
Nab2 Polyclonal specifically detects Nab2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Nab2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| EGR-1-binding protein 2, MADEREGR1 binding protein 2, Melanoma-associated delayed early response protein, MGC75085, NGFI-A binding protein 2 (EGR1 binding protein 2), NGFI-A-binding protein 2, Protein MADER | |
| Rabbit | |
| 57 kDa | |
| 0.1 mL | |
| Neuroscience, Transcription Factors and Regulators | |
| 4665 | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunoprecipitation 1:100-1:500 | |
| Q15742 | |
| NAB2 | |
| Recombinant protein encompassing a sequence within the center region of human NAB2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction