missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAB2 Monoclonal Antibody (OTI1B10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804061
This item is not returnable.
View return policy
Description
This gene encodes a member of the family of NGFI-A binding (NAB) proteins, which function in the nucleus to repress transcription induced by some members of the EGR (early growth response) family of transactivators. NAB proteins can homo- or hetero-multimerize with other EGR or NAB proteins through a conserved N-terminal domain, and repress transcription through two partially redundant C-terminal domains. Transcriptional repression by the encoded protein is mediated in part by interactions with the nucleosome remodeling and deactylase (NuRD) complex. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Specifications
| NAB2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15742 | |
| NAB2 | |
| Human recombit protein fragment corresponding to amino acids 178-442 of human NAB2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI1B10 | |
| Unconjugated | |
| NAB2 | |
| MADER | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4665 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction