Learn More
NAB2 Monoclonal Antibody (OTI1B10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA804061S
Encompass_Preferred
 View more versions of this product
Product Code. 17593403
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
NAB2 Monoclonal antibody specifically detects NAB2 in Human samples. It is validated for Western Blot
Specifications
Specifications
NAB2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q15742
Mouse
Affinity chromatography
RUO
4665
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Western Blot
OTI1B10
Unconjugated
NAB2
AI451907; EGR1 binding protein 2; EGR-1-binding protein 2; MADER; Melanoma-associated delayed early response protein; NAB2; NGFI-A binding protein 2; NGFI-A binding protein 2 (EGR1 binding protein 2); NGFI-A binding protein 2 (EGR-1 binding protein 2); NGFI-A-binding protein 2; Protein MADER
Human recombinant protein fragment corresponding to amino acids 178-442 of human NAB2 produced in E.coli.
30 μL