missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAB2 Monoclonal Antibody (OTI3H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803957S
This item is not returnable.
View return policy
Description
NAB2 Monoclonal antibody specifically detects NAB2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| NAB2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15742 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4665 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3H5 | |
| Unconjugated | |
| NAB2 | |
| AI451907; EGR1 binding protein 2; EGR-1-binding protein 2; MADER; Melanoma-associated delayed early response protein; NAB2; NGFI-A binding protein 2; NGFI-A binding protein 2 (EGR1 binding protein 2); NGFI-A binding protein 2 (EGR-1 binding protein 2); NGFI-A-binding protein 2; Protein MADER | |
| Human recombinant protein fragment corresponding to amino acids 178-442 of human NAB2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction