missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NABP1 Mouse anti-Human, Clone: OTI6B10, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811088S
This item is not returnable.
View return policy
Description
NABP1 Monoclonal antibody specifically detects NABP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| NABP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96AH0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 64859 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6B10 | |
| Unconjugated | |
| NABP1 | |
| 4930434H03Rik; 4930442A21Rik; 4930488J04Rik; 4933440J18Rik; 5830411E10Rik; AI852561; hSSB2; Nabp1; Nbp1; nucleic acid binding protein 1; nucleic acid-binding protein 1; Obfc2a; oligonucleotide/oligosaccharide-binding fold containing 2A; oligonucleotide/oligosaccharide-binding fold-containing protein 2A; RGD1306658; Sensor of single-strand DNA complex subunit B2; sensor of ssDNA subunit B2; Single-stranded DNA-binding protein 2; SOSS complex subunit B2; SOSS-B2; Ssb2 | |
| Full length human recombinant protein of human NABP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction