missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAC1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98709UV
This item is not returnable.
View return policy
Description
NAC1 Polyclonal specifically detects NAC1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| NAC1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| BEND8, BTB (POZ) domain containing 14B, BTB/POZ domain-containing protein 14B, BTBD14B, NAC1BEN domain containing 8, NAC-1FLJ37383, nucleus accumbens associated 1, BEN and BTB (POZ) domain containing, nucleus accumbens-associated protein 1, transcriptional repressor NAC1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human NAC1. | |
| 0.1 mL | |
| Cancer, Chromatin Research, Stem Cell Markers, Transcription Factors and Regulators | |
| 112939 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction