missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NACA Polyclonal Antibody
Description
NACA Polyclonal Antibody for Western Blot
NACA prevents inappropriate targeting of non-secretory polypeptides to the endoplasmic reticulum (ER). It binds to nascent polypeptide chains as they emerge from the ribosome and blocks their interaction with the signal recognition particle (SRP), which normally targets nascent secretory peptides to the ER. NACA also reduces the inherent affinity of ribosomes for protein translocation sites in the ER membrane (M sites), and may act as a specific coactivator for JUN, binding to DNA and stabilizing the interaction of JUN homodimers with target gene promoters.
Specifications
Specifications
| Antigen | NACA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | NACA |
| Gene Accession No. | E9PAV3, P70670, Q13765, Q60817 |
| Gene Alias | AL022831; AL024382; Alpha-NAC; Alpha-NAC, muscle-specific form; alpha-NAC/1.9.2; Gm1878; Hom s 2; HSD48; mKIAA0363; NACA; NACA1; NAC-alpha; nascent polypeptide-associated complex alpha polypeptide; nascent polypeptide-associated complex alpha subunit; nascent polypeptide-associated complex subunit alpha; Nascent polypeptide-associated complex subunit alpha, muscle-specific form; nascent-polypeptide-associated complex alpha polypeptide; skNAC |
| Gene Symbols | NACA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?