missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford NADH
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Substance Name: B-Nicotinamide adenine dinucleotide, reduced form, disodium salt ,CAS Number: 606-68-8 , Molecular Formula: C21H27N7O14 P2Na2 : 3H2O , Molecular Weight: 763.5 , Chemical Purity: >98%, Soluble in: Water , Store at: 2°C to 8°C
Specifications
Specifications
| Quantity | 1 g |
| Molecular Formula | C21H27N7O14P2Na2-3H2O |
| CAS | 606-68-8 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction