missing translation for 'onlineSavingsMsg'
Learn More

NADH disodium salt, MedChemExpress

Product Code. p-200101533
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16442326

Brand: Medchem Express HYF0001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NADH disodium salt (Disodium NADH) is an orally active reduced coenzyme. NADH disodium salt is a donor of ADP-ribose units in ADP-ribosylaton reactions and a precursor of cyclic ADP-ribose. NADH disodium salt plays a role as a regenerative electron donor in cellular energy metabolism, including glycolysis, β-oxidation and the tricarboxylic acid (TCA) cycle.

NADH disodium salt (Disodium NADH) is an orally active reduced coenzyme. NADH disodium salt is a donor of ADP-ribose units in ADP-ribosylaton reactions and a precursor of cyclic ADP-ribose. NADH disodium salt plays a role as a regenerative electron donor in cellular energy metabolism, including glycolysis, β-oxidation and the tricarboxylic acid (TCA) cycle.

Specifications

Chemical Name or Material NADH disodium salt
CAS 606-68-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H27N7Na2O14P2
Quantity 500 mg
Synonym Disodium NADH
Solubility Information H2O : ≥ 100 mg/mL (140.96 mM) ∣DMSO : 100 mg/mL (140.96 mM; Need ultrasonic)
SMILES NC1=C2C(N([C@H]3[C@H](O)[C@H](O)[C@@H](COP(OP([O-])(OC[C@@H]4[C@@H](O)[C@@H](O)[C@H](N5C=C(C(N)=O)CC=C5)O4)=O)([O-])=O)O3)C=N2)=NC=N1.[Na+].[Na+]
Molecular Weight (g/mol) 709.4
Formula Weight 709.4
Percent Purity 95.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.