Learn More
NAE1 Mouse anti-Human, Clone: OTI1D12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF804320
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene binds to the beta-amyloid precursor protein. Beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer′s disease. In addition, the encoded protein can form a heterodimer with UBE1C and bind and activate NEDD8, a ubiquitin-like protein. This protein is required for cell cycle progression through the S/M checkpoint. Three transcript variants encoding different isoforms have been found for this gene.Specifications
| NAE1 | |
| Monoclonal | |
| Unconjugated | |
| NAE1 | |
| A-116A10.1, APPBP1, HPP1, ula-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8883 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1D12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q13564 | |
| NAE1 | |
| Human recombit protein fragment corresponding to amino acids 1-274 of human NAE1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.