missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAE1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16548161
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16548161 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16548161 Supplier Invitrogen™ Supplier No. PA592174

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MAQLGKLLKE QKYDRQLRLW GDHGQEALES AHVCLINATA TGTEILKNLV LPGIGSFTII DGNQVSGEDA GNNFFLQRSS IGKNRAEAAM EFLQELNSDV SGSFVEESPE NLLDNDPSFF CRFTVVVATQ LPESTSLRLA DVLWNSQIPL LICRTYGLVG YMRIIIKEHP VIESHPDNAL EDLRLDKPFP ELREHFQSYD LDHMEKKDHS HTPWIVIIAK; Positive Samples: SH-SY5Y, BT-474, HL-60, SW620, NIH/3T3, Mouse brain, Mouse lung, Rat brain; Cellular Location: Cell membrane.

The protein encoded by this gene binds to the beta-amyloid precursor protein. Beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer's disease. In addition, the encoded protein can form a heterodimer with UBE1C and bind and activate NEDD8, a ubiquitin-like protein. This protein is required for cell cycle progression through the S/M checkpoint. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAE1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene NAE1
Gene Accession No. Q13564, Q8VBW6, Q9Z1A5
Gene Alias 59kDa; A-116A10.1; amyloid beta precursor protein binding protein 1, 59kDa; amyloid beta precursor protein-binding protein 1, 59 kDa; amyloid beta precursor protein-binding protein 1, 59kD; amyloid protein-binding protein 1; Appbp1; APP-BP1; HPP1; Nae1; NEDD8 activating enzyme E1 subunit 1; NEDD8-activating enzyme E1 regulatory subunit; NEDD8-activating enzyme E1 subunit; protooncogene protein 1; proto-oncogene protein 1; ula-1
Gene Symbols NAE1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human NAE1 (NP_0038961).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234664, 84019, 8883
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.