missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAF1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22263FR
This item is not returnable.
View return policy
Description
NAF1 Polyclonal antibody specifically detects NAF1 in Human samples. It is validated for Western Blot
Specifications
| NAF1 | |
| Polyclonal | |
| Western Blot | |
| nuclear assembly factor 1 homolog (S. cerevisiae) | |
| The immunogen this antibody was made to, maps to a region between residue 50 and 100 of human Nuclear Assembly Factor 1 using the numbering given in entry NP_612395.2 (GeneID 92345). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 92345 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction