missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAGA Monoclonal Antibody (OTI7F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811277
This item is not returnable.
View return policy
Description
NAGA (N-acetylgalactosaminidase, alpha), also known as alpha-galactosidase B or GALB, is a 411 lysosomal protein belonging to the glycosyl hydrolase 27 family that may exist as a homodimer and plays a critical role in glycolipid breakdown. NAGA encodes alpha-N-acetylgalactosaminidase, a lysosomal enzyme, which cleaves alpha-N-acetylgalactosaminyl groups from glycoconjugates. Mapping to human chromosome 22q13.2, NAGA defects are the cause of an autosomal recessive disorder with three phenotypes, known as Schindler disease (types I, II and III) or NAGA deficiency (types I, II and III). Characterized by neurologic manifestations that range in severity, Schindler disease type I is the most severe form, followed by type III, which may have mild-to-moderate effects. Schindler disease type II, also known as Kanzaki disease, is characterized by mild intellectual impairment and angiokeratoma corporis diffusum.Specifications
| NAGA | |
| Monoclonal | |
| Unconjugated | |
| NAGA | |
| D22S674, GALB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4668 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI7F1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P17050 | |
| NAGA | |
| Full length human recombit protein of human NAGA produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction