missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAGLU Polyclonal Antibody
GREENER_CHOICE

Product Code. 17750704
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17750704 200 μL 200µL
17773144 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17750704 Supplier Invitrogen™ Supplier No. PA5119299

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NAGLU Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes an enzyme that degrades heparan sulfate by hydrolysis of terminal N-acetyl-D-glucosamine residues in N-acetyl-alpha-D-glucosaminides. Defects in this gene are the cause of mucopolysaccharidosis type IIIB (MPS-IIIB), also known as Sanfilippo syndrome B. This disease is characterized by the lysosomal accumulation and urinary excretion of heparan sulfate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAGLU
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene NAGLU
Gene Accession No. P54802
Gene Alias Alpha-N-acetylglucosaminidase; alpha-N-acetylglucosaminidase (Sanfilippo disease IIIB); Alpha-N-acetylglucosaminidase 77 kDa form; Alpha-N-acetylglucosaminidase 82 kDa form; alpha-N-acetylglucosaminidase, lysosomal; CMT2V; MPS3B; MPS-IIIB; N-acetyl-alpha-glucosaminidase; N-acetyl-glucosaminidase; N-acetylglucosaminidase, alpha; NAG; NAGLU; RGD1564228; testicular tissue protein Li 18; UFHSD; UFHSD1
Gene Symbols NAGLU
Host Species Rabbit
Immunogen Recombinant protein N-Acetyl Alpha-D-Glucosaminidase. The antigen corresponds to amino acid range 485-743 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100519685, 4669
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.