missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAGLU Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558551
This item is not returnable.
View return policy
Description
Immunogen sequence: LQGWLFQHQP QFWGPAQIRA VLGAVPRGRL LVLDLFAESQ PVYTRTASFQ GQPFIWCMLH NFGGNHGLFG ALEAVNGGPE AARLFPNSTM VGTGMAPEGI SQNEVVYSLM A Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
This gene encodes an enzyme that degrades heparan sulfate by hydrolysis of terminal N-acetyl-D-glucosamine residues in N-acetyl-alpha-D-glucosaminides. Defects in this gene are the cause of mucopolysaccharidosis type IIIB (MPS-IIIB), also known as Sanfilippo syndrome B. This disease is characterized by the lysosomal accumulation and urinary excretion of heparan sulfate.
Specifications
| NAGLU | |
| Polyclonal | |
| Unconjugated | |
| NAGLU | |
| Alpha-N-acetylglucosaminidase; alpha-N-acetylglucosaminidase (Sanfilippo disease IIIB); Alpha-N-acetylglucosaminidase 77 kDa form; Alpha-N-acetylglucosaminidase 82 kDa form; alpha-N-acetylglucosaminidase, lysosomal; CMT2V; MPS3B; MPS-IIIB; N-acetyl-alpha-glucosaminidase; N-acetyl-glucosaminidase; N-acetylglucosaminidase, alpha; NAG; NAGLU; RGD1564228; testicular tissue protein Li 18; UFHSD; UFHSD1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27419, 4669 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P54802 | |
| NAGLU | |
| Recombinant protein corresponding to Human NAGLU. Recombinant protein control fragment (Product #RP-93204). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction